Learn More
Invitrogen™ ULBP1 Monoclonal Antibody (170818)

Description
This antibody stains human ULBP-1 cell transfectants. This antibody does not stain cells transfected with ULBP-2 or ULBP-3. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | ULBP1 |
| Applications | Flow Cytometry, Neutralization |
| Classification | Monoclonal |
| Clone | 170818 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | ULBP1 |
| Gene Accession No. | Q9BZM6 |
| Gene Alias | A430108B07Rik; ALCAN-beta; MULT1; N2DL1; N2DL-1; NKG2D ligand; NKG2D ligand 1; NKG2D ligand1; NKG2DL1; RAET1I; retinoic acid early transcript 1I; UL16 binding protein 1; UL16-binding protein 1; UL16-binding protein-like transcript 1; ULBP; ULBP 1; ULBP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.