Learn More
Invitrogen™ ULBP2 Recombinant Rabbit Monoclonal Antibody (003)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529880
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human ULBP2.
ULBP2 encodes a major histocompatibility complex (MHC) class I-related molecule that binds to the NKG2D receptor on natural killer (NK) cells to trigger release of multiple cytokines and chemokines that in turn contribute to the recruitment and activation of NK cells. The encoded protein undergoes further processing to generate the mature protein that is either anchored to membrane via a glycosylphosphatidylinositol moiety, or secreted. Many malignant cells secrete the encoded protein to evade immunosurveillance by NK cells. This gene is located in a cluster of multiple MHC class I-related genes on chromosome 6.
Specifications
ULBP2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9BZM5 | |
ULBP2 | |
Recombinant Human ULBP2 protein (Met1-Ser217). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
003 | |
Unconjugated | |
ULBP2 | |
ALCAN-alpha; N2DL2; N2DL-2; NKG2D ligand 2; NKG2DL2; RAET1H; Retinoic acid early transcript 1H; UL16 binding protein 2; UL16-binding protein 2; ULBP2; UNQ463/PRO791 | |
Rabbit | |
Protein A | |
RUO | |
80328 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.