Learn More
Invitrogen™ ULBP2/ULBP5/ULBP6 Monoclonal Antibody (165903)

Description
This antibody detects human ULBP-2 and human RAET1L/ULBP-6 in direct ELISA and stains cells transfected with human ULBP-2, human ULBP-5 or human RAET1L/ULBP-6 in flow cytometry. In direct ELISA, 11% cross-reactivity with recombinant human ULBP5 is observed. This antibody does not stain cells transfected with human ULBP-1 or human ULBP-3 in flow cytometry. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
Specifications
| Antigen | ULBP2/ULBP5/ULBP6 |
| Applications | Flow Cytometry, Neutralization |
| Classification | Monoclonal |
| Clone | 165903 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | RAET1G |
| Gene Accession No. | Q6H3X3, Q9BZM5 |
| Gene Alias | ALCAN-alpha; N2DL2; N2DL-2; NKG2D ligand 2; NKG2DL2; RAET1G; RAET1H; retinoic acid early transcript 1G; retinoic acid early transcript 1G protein; Retinoic acid early transcript 1H; UL16 binding protein 2; UL-16 binding protein 5; UL16-binding protein 2; ULBP; ULBP2; ULBP5; UNQ463/PRO791 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.