Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ulex Europaeus (Gorse) Agglutinin I (UEA I), fluorescein (FITC)

Catalog No. L32476
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L32476 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L32476 Supplier Invitrogen™ Supplier No. L32476
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Ulex Europaeus Agglutinin I (UEA I), fluorescein (FITC), is a bright-green UEA I-fluorophore conjugate.

UEA I interacts specifically with blood group H type 2 antigen, which is expressed in individuals with type O blood and to a lesser degree in individuals with type A or B blood. UEA I has been used to differentiate between angiosarcomas and epithelial tumors. However, blood group antigens are found in a variety of tissues, so better results may be obtained with the use of specific monoclonal antibodies. UEA I is also a marker for incompletely differentiated gastrin cells.

• Very bright FITC-conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
• Applications: immunofluorescence, glycobiology
• Sugar specificity: fucose, arabinose
• Inhibiting/eluting sugar: 50–100 mM L-fucose
• Blood group specificity: O (H)
• Unconjugated lectin molecular weight: 63 KDa
• Immunofluorescence working concentration: 5–20 μg/mL

UEA I-FITC is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2+.

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Fluorescein
Quantity 2 mg
Source Gorse, furze
Content And Storage When stored at or below 2°C to 8°C in the dark, the product is stable for 6 months after receipt.

Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein Family Lectins

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.