Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ulex Europaeus Lectin 1 Native Protein
SDP

Catalog No. NB398374 Shop All Novus Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB398374 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB398374 Supplier R&D Systems™ Supplier No. NBP2427005MG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Native Protein

Agglutination activity is expressed in μg/mL and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group O erythrocytes after 1 hour incubation at 25 degrees C.

Specifications

For Use With (Application) SDS-PAGE, Bioactivity
Formulation Lyophilized in buffer. Information about buffer components is properietary.
Molecular Weight (g/mol) MolecularWeight-theroretical: 68 kDa
Quantity 5 mg
Storage Requirements Store at 4°C. Do not freeze.
Gene Alias Gorse or Furze Lectin, UEA-1; H(O) lectin 1; Anti H(O) lectin I
Conjugate Unconjugated
Species Plants
Recombinant Native
Reconstitution Reconstitute with PBS (pH 6.8) to a final concentration of 1 mg/mL
Inhibitor Properties Ulex europaeus agglutinin has anti-H blood group specificity.
Form Purified
Product Line Novus
Purity or Quality Grade >50% SDS-PAGE
Protein Ulex Europaeus Lectin 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.