Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULK1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$196.10 - $499.50
Specifications
Antigen | ULK1 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
ULK1 Polyclonal specifically detects ULK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
ULK1 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
ATG1, ATG1 autophagy related 1 homolog, ATG1A, EC 2.7.11, EC 2.7.11.1, FLJ38455, FLJ46475, KIAA0722, serine/threonine-protein kinase ULK1, UNC51, unc-51 (C. elegans)-like kinase 1, Unc51.1, Unc-51-like kinase 1, unc-51-like kinase 1 (C. elegans) | |
ULK1 | |
IgG | |
Affinity Purified | |
105 kDa |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Autophagy, mTOR Pathway, Protein Kinase | |
PBS with 0.02% Sodium Azide | |
8408 | |
Antibody was raised against a 16 amino acid peptide near the center of human ULK1 . The immunogen is located within amino acids 350 - 400 of ULK1. Amino Acid Squence: CDLVAEAPSAKPPPDSL | |
Primary | |
Two alternatively spliced transcript variants encoding different isoforms have been identified. ULK1 antibody is predicted to not cross-react with ULK2. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title