Learn More
Promega ULK1 Kinase Enzyme System, 1mg
Catalog No. PRV3522 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3522 1 mg
1 options
Catalog No. PRV3522 Supplier Promega Supplier No. V3522
Add to Cart
Add to Cart
Request a Quote
Recombinant human ULK1 (1-649) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ULK1 is a serine/threonine protein kinase that plays a critical role during initial stages of autophagy, a vital response to nutrient starvation.
  • Easily Screen and Profile ULK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C