Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ULK1, p Ser556 Antibody (SR1119), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322612100UL
Description
ULK1 Monoclonal antibody specifically detects ULK1 in Human, Mouse samples. It is validated for Western BlotSpecifications
ULK1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
SR1119 | |
Western Blot 1:500-1:2000 | |
ATG1, ATG1 autophagy related 1 homolog, ATG1A, EC 2.7.11, EC 2.7.11.1, FLJ38455, FLJ46475, KIAA0722, serine/threonine-protein kinase ULK1, UNC51, unc-51 (C. elegans)-like kinase 1, Unc51.1, Unc-51-like kinase 1, unc-51-like kinase 1 (C. elegans) | |
A synthesized peptide derived from human phospho-ULK1 [Ser556] (Uniprot #: O75385) | |
100 μg | |
Autophagy, mTOR Pathway, Protein Kinase | |
8408 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction