Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULK1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 209861AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-3191


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

ULK1, also known as ATG1, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK1 results in a severe defect in the autophagy pathway. ULK1 is highly conserved among eukaryotes, and are the Unc-51-like kinases, ULK1 and ULK2 in mammals. ULK1 is ubiquitously expressed and involved in autophagy in response to starvation. It is the target of the TOR kinase signaling pathway that regulates autophagy through the control of phosphorylation status of ATG13. ULK1 also plays a role early in neuronal differentiation.
Specifications

Specifications

ULK1
Polyclonal
Unconjugated
ULK1
ATG1, KIAA0722, ULK1, Unc 51 like kinase 1, UNC51, Unc51.1
Rabbit
Antigen Affinity Chromatography
RUO
360827, 8408
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75385
Ulk1
Peptide
150 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.