Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ULK1/ULK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116888 Supplier Invitrogen™ Supplier No. PA5116888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: RD, SH-SY5Y, HepG2 Immunogen sequence: MEPGRGGTET VGKFEFSRKD LIGHGAFAVV FKGRHREKHD LEVAVKCINK KNLAKSQTLL GKEIKILKEL KHENIVALYD FQEMANSVYL VMEYCNGGDL ADYLHAMRTL SEDTIRLFLQ QIAGAMRLLH SKGIIHRDLK PQNILLSNPA GRRANPNSIR VKIADFGFAR YLQSNMMAAT LCGSPMYMAP EVIMSQHYDG KADLWSIGTI VYQCLTGKAP FQASSPQDLR LFYEKNKTLV PTIPRETSAP LRQLLLALLQ RNHKDRMDFD EFFHHPFLDA SPS.

Mammalian ULK1 (unc-51 like kinase 1) and ULK2, Caenorhabditis elegans UNC-51, and Drosophila melanogaster Atg1 are serine/threonine kinases that regulate flux through the autophagy pathway in response to various types of cellular stress.ULK1 and ULK2 all have different tissue-specific expressions partially contributing to diverse and ULK-specific interaction networks in various tissues.ULK1 (or its close family member ULK2) interacts with several other proteins to form a large complex that is required for the formation of an autophagosome. The other proteins that complex with ULK1/ULK2 include ATG13, ATG101, and RB1CC1 (FIP200/ATG17).AMPK, activated during low nutrient conditions, directly phophorylates ULK1 at multiple sites including Ser317, Ser555, and Ser777. Conversely, mTOR, which is a regulator of cell growth and is an inhibitor of autophagy, phosphorylates ULK1 at Ser757 and disrupts the interaction between ULK1 and AMPK.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULK1/ULK2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ULK1
Gene Accession No. O75385, Q8IYT8
Gene Alias ATG1; ATG1 autophagy related 1 homolog; ATG1A; ATG1B; Autophagy-related protein 1 homolog; hATG1; KIAA0623; KIAA0722; Serine/threonine-protein kinase ULK1; serine/threonine-protein kinase ULK2; ULK1; ULK2; UNC51; unc-51 like autophagy activating kinase 1; unc-51 like autophagy activating kinase 2; Unc51.1; Unc51.2; Unc-51-like kinase 1; Unc-51-like kinase 2
Gene Symbols ULK1, ULK2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-283 of human ULK1/ULK2 (NP_0035561).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8408, 9706
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.