Learn More
ULK2 Polyclonal Antibody, Invitrogen™
Description
Specifications
Specifications
| Antigen | ULK2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | ULK2 |
| Gene Accession No. | Q8IYT8, Q9QY01 |
| Gene Alias | A830085I22Rik; ATG1B; AU015340; EC:2.7.11.1; KIAA0623; mKIAA0623; serine/threonine-protein kinase ULK2; Serine/threonine-protein kinase Unc51.2; ULK2; unc-51 like autophagy activating kinase 2; unc-51 like kinase 2; Unc51.2; unc-51-like kinase 2 |
| Gene Symbols | ULK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.