Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ULK2 Recombinant Rabbit Monoclonal Antibody (HL2206)
GREENER_CHOICE

Catalog No. FEMA550432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550432 Supplier Invitrogen™ Supplier No. MA550432
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

ULK2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

ULK2, also known as ATG1B, is a key serine/threonine protein kinase probably acting at the most upstream step of autophagosome formation. Knockout of ULK2 results in a severe defect in the autophagy pathway. ULK2 is highly conserved among eukaryotes and shows high homology with its related protein ULK1. Both ULK1 and ULK2 form a complex with ATG13 and FIP200 that mediates TOR signaling and is essential for autophagy. Like ULK1, ULK2 is also thought to be involved in early neuronal growth and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULK2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2206
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ULK2
Gene Accession No. Q8IYT8
Gene Alias A830085I22Rik; ATG1B; AU015340; EC:2.7.11.1; KIAA0623; mKIAA0623; serine/threonine-protein kinase ULK2; Serine/threonine-protein kinase Unc51.2; ULK2; unc-51 like autophagy activating kinase 2; unc-51 like kinase 2; Unc51.2; unc-51-like kinase 2
Gene Symbols ULK2
Host Species Rabbit
Immunogen Recombinant fragmemt of human ULK2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303206, 9706
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.