Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ULK3 Monoclonal Antibody (1404CT148.89.25)
GREENER_CHOICE

Catalog No. PIMA537660
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537660 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537660 Supplier Invitrogen™ Supplier No. MA537660
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ULK3 Monoclonal Antibody for Western Blot, IHC (P)

ULK3 or unc-51-like kinase 3 is a member of the serine/threonine kinase family that is involved in the SHH signaling pathway as a positive regulator of GLI proteins. ULK3 enhances endogenous and over-expressed GLI transcriptional activity in cultured cells and alters its subcellular localization. Furthermore, ULK3 phosphorylates GLI proteins in vitro. ULK3 is widely expressed and its expression is higher in a number of tissues where SHH signaling is known to be active, again suggesting that ULK3 is involved in the SHH pathway as a positive regulator of GLI proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULK3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1404CT148.89.25
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ULK3
Gene Accession No. D3ZHP7, Q3U3Q1, Q6PHR2
Gene Alias 1200015E14Rik; DKFZP434C131; EC:2.7.11.1; FLJ90566; Serine/threonine-protein kinase ULK3; ULK3; unc-51 like kinase 3; unc-51-like kinase 3
Gene Symbols ULK3
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 435-468 amino acids from the C-terminal region of human ULK3.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25989, 691171, 71742
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.