Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ULK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA515397
Description
This antibody is predicted to react with mouse based on sequence homology.
ULK3 or unc-51-like kinase 3 is a member of the serine/threonine kinase family that is involved in the SHH signaling pathway as a positive regulator of GLI proteins. ULK3 enhances endogenous and over-expressed GLI transcriptional activity in cultured cells and alters its subcellular localization. Furthermore, ULK3 phosphorylates GLI proteins in vitro. ULK3 is widely expressed and its expression is higher in a number of tissues where SHH signaling is known to be active, again suggesting that ULK3 is involved in the SHH pathway as a positive regulator of GLI proteins.
Specifications
ULK3 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q3U3Q1, Q6PHR2 | |
ULK3 | |
KLH conjugated synthetic peptide between 14-43 amino acids from the N-terminal region of human ULK3. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ULK3 | |
1200015E14Rik; DKFZP434C131; EC:2.7.11.1; FLJ90566; Serine/threonine-protein kinase ULK3; ULK3; unc-51 like kinase 3; unc-51-like kinase 3 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
25989, 71742 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction