Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ULK3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA559193

Catalog No. PIPA559193


Only null left
Add to Cart

Description

Description

Immunogen sequence: ILKGIRHPHI VQLKDFQWDS DNIYLIMEFC AGGDLSRFIH TRRILPEKVA RVFMQQLASA LQFLHERNIS HLDLKPQNIL LSSLEKPHLK LADFGFAQHM SPWDEKHVLR GSPLYMAPEM VCQRQYDARV DLWSMGVILY GETSFPC Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.

ULK3 or unc-51-like kinase 3 is a member of the serine/threonine kinase family that is involved in the SHH signaling pathway as a positive regulator of GLI proteins. ULK3 enhances endogenous and over-expressed GLI transcriptional activity in cultured cells and alters its subcellular localization. Furthermore, ULK3 phosphorylates GLI proteins in vitro. ULK3 is widely expressed and its expression is higher in a number of tissues where SHH signaling is known to be active, again suggesting that ULK3 is involved in the SHH pathway as a positive regulator of GLI proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ULK3
Polyclonal
Unconjugated
ULK3
1200015E14Rik; DKFZP434C131; EC:2.7.11.1; FLJ90566; Serine/threonine-protein kinase ULK3; ULK3; unc-51 like kinase 3; unc-51-like kinase 3
Rabbit
Antigen Affinity Chromatography
RUO
25989, 691171, 71742
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
D3ZHP7, Q3U3Q1, Q6PHR2
ULK3
Recombinant protein corresponding to Human ULK3. Recombinant protein control fragment (Product #RP-89789).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.