Learn More
Invitrogen™ ULK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580207
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Testis Tissue, Mouse Brain Tissue, Human Placenta Tissue, 22RV1 whole cell, SMMC whole cell. IHC: rat brain tissue, human lung cancer tissue. Flow: U20S cell, THP-1 cell.
ULK3 or unc-51-like kinase 3 is a member of the serine/threonine kinase family that is involved in the SHH signaling pathway as a positive regulator of GLI proteins. ULK3 enhances endogenous and over-expressed GLI transcriptional activity in cultured cells and alters its subcellular localization. Furthermore, ULK3 phosphorylates GLI proteins in vitro. ULK3 is widely expressed and its expression is higher in a number of tissues where SHH signaling is known to be active, again suggesting that ULK3 is involved in the SHH pathway as a positive regulator of GLI proteins.
Specifications
ULK3 | |
Polyclonal | |
Unconjugated | |
ULK3 | |
1200015E14Rik; DKFZP434C131; EC:2.7.11.1; FLJ90566; Serine/threonine-protein kinase ULK3; ULK3; unc-51 like kinase 3; unc-51-like kinase 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25989, 691171, 71742 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 5mg BSA and 0.05mg sodium azide | |
D3ZHP7, Q3U3Q1, Q6PHR2 | |
ULK3 | |
A synthetic peptide corresponding to a sequence at C-terminus of human ULK3 (427-454aa HTEVQNLMARAEYLKEQVKMRESRWEAD). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.