Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULK3 Rabbit anti-Bovine, Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502209587
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9587 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9587 Supplier Fabgennix Inc Supplier No. ULK3301AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ULK3 Polyclonal antibody specifically detects ULK3 in Bovine, Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen ULK3
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ULK3
Gene Accession No. D3ZHP7, Q3U3Q1, Q6PHR2
Gene Alias 1200015E14Rik; DKFZP434C131; EC:2.7.11.1; FLJ90566; Serine/threonine-protein kinase ULK3; ULK3; unc-51 like kinase 3; unc-51-like kinase 3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within C terminal amino acids 452-472 of Mouse ULK3, coupled to a carrier protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25989, 453747, 506063, 691171, 71742
Target Species Bovine, Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.