Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ULK3 Recombinant Rabbit Monoclonal Antibody (5Z7Z6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542879
This item is not returnable.
View return policy
Description
Immunogen sequence: KPRLLAALEV ASAAMAKEEA AGGEQDALDL YQHSLGELLL LLAAEPPGRR RELLHTEVQN LMARAEYLKE QVKMRESRWE ADTLDKEGLS ESVRSSCTLQ.
ULK3 or unc-51-like kinase 3 is a member of the serine/threonine kinase family that is involved in the SHH signaling pathway as a positive regulator of GLI proteins. ULK3 enhances endogenous and over-expressed GLI transcriptional activity in cultured cells and alters its subcellular localization. Furthermore, ULK3 phosphorylates GLI proteins in vitro. ULK3 is widely expressed and its expression is higher in a number of tissues where SHH signaling is known to be active, again suggesting that ULK3 is involved in the SHH pathway as a positive regulator of GLI proteins.
Specifications
ULK3 | |
Recombinant Monoclonal | |
0.7 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D3ZHP7, Q3U3Q1, Q6PHR2 | |
ULK3 | |
A synthetic peptide corresponding to a sequence within amino acids 373-472 of human ULK3 (Q6PHR2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot, Immunocytochemistry | |
5Z7Z6 | |
Unconjugated | |
ULK3 | |
1200015E14Rik; DKFZP434C131; EC:2.7.11.1; FLJ90566; Serine/threonine-protein kinase ULK3; ULK3; unc-51 like kinase 3; unc-51-like kinase 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25989, 691171, 71742 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction