Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ULK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA538038
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538038 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538038 Supplier Invitrogen™ Supplier No. PA538038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A recommended positive control is NIH-3T3 cell lysate. Multiple isoforms of ULK4 are known to exist.

The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. ULK4 (unc-51-like kinase 4) is a 1,275 amino acid protein that contains one protein kinase domain and one HEAT repeat and belongs to the Ser/Thr protein kinase family. Although containing what is thought to be a catalytically inactive domain, ULK4 may play a role in the ATP-dependent phosphorylation of target proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULK4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ULK4
Gene Accession No. Q3V129, Q96C45
Gene Alias 4932415A06Rik; A730098P15; FAM7C1; REC01035; RGD1565726; serine/threonine-protein kinase ULK4; ULK4; unc-51 like kinase 4; unc-51-like kinase 4
Gene Symbols ULK4
Host Species Rabbit
Immunogen 16 amino acid peptide near the carboxy terminus of human ULK4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209012, 54986
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.