Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ULK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553672
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553672 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553672 Supplier Invitrogen™ Supplier No. PA553672
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LTREIKHKNI VTFHEWYETS NHLWLVVELC TGGSLKTVIA QDENLPEDVV REFGIDLISG LHHLHKLGIL FCDISPRKIL LEGPGTLKFS NFCLAKVEGE NLEEFFALVA AEEGGGDNGE NVLKKSMKSR VKGSPVYTAP EV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 30%.

The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. ULK4 (unc-51-like kinase 4) is a 1,275 amino acid protein that contains one protein kinase domain and one HEAT repeat and belongs to the Ser/Thr protein kinase family. Although containing what is thought to be a catalytically inactive domain, ULK4 may play a role in the ATP-dependent phosphorylation of target proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ULK4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ULK4
Gene Accession No. Q96C45
Gene Alias 4932415A06Rik; A730098P15; FAM7C1; REC01035; RGD1565726; serine/threonine-protein kinase ULK4; ULK4; unc-51 like kinase 4; unc-51-like kinase 4
Gene Symbols ULK4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ULK4. Recombinant protein control fragment (Product #RP-91436).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54986
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.