Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Ultrafiltration Membrane Discs

Catalog No. 14558230 Shop All Sartorius Products
Change view
Click to view available options
Diameter (Metric):
25 mm
47 mm
63 mm
76 mm
150 mm
Material (Membrane):
Cellulose
PES
Polysulfone
Thickness (Metric):
120 μm
180 μm
MWCO:
1 kDa
10 kDa
100 kDa
30 kDa
300 kDa
5 kDa
50 kDa
32 product options available for selection
Product selection table with 32 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Material (Membrane) Thickness (Metric) MWCO
14-558-230 25 mm Cellulose 180 μm 10 kDa
14-558-242 25 mm Polysulfone 120 μm 5 kDa
14-558-255 76 mm Polysulfone 120 μm 50 kDa
14-558-225 25 mm Cellulose 180 μm 5 kDa
14-558-245 76 mm Polysulfone 120 μm 5 kDa
14-558-257 47 mm Polysulfone 120 μm 30 kDa
14-558-251 76 mm Polysulfone 120 μm 10 kDa
14-558-234 25 mm Cellulose 180 μm 30 kDa
14-558-244 63 mm Polysulfone 120 μm 5 kDa
14-558-228 63 mm Cellulose 180 μm 5 kDa
14-558-247 25 mm Polysulfone 120 μm 10 kDa
14-558-260 47 mm PES 120 μm 100 kDa
14-558-239 47 mm Polysulfone 120 μm 1 kDa
14-558-258 63 mm Polysulfone 120 μm 30 kDa
14-558-227 47 mm Cellulose 180 μm 5 kDa
14-558-259 76 mm Polysulfone 120 μm 30 kDa
14-558-246 150 mm Polysulfone 120 μm 10 kDa
14-558-253 47 mm Polysulfone 120 μm 50 kDa
14-558-237 76 mm Cellulose 180 μm 30 kDa
14-558-233 76 mm Cellulose 180 μm 10 kDa
14-558-235 47 mm Cellulose 180 μm 30 kDa
14-558-264 76 mm Polysulfone 120 μm 300 kDa
14-558-236 63 mm Cellulose 180 μm 30 kDa
14-558-240 63 mm Polysulfone 120 μm 1 kDa
14-558-249 47 mm Polysulfone 120 μm 10 kDa
14-558-229 76 mm Cellulose 180 μm 5 kDa
14-558-262 47 mm Polysulfone 120 μm 300 kDa
14-558-241 76 mm Polysulfone 120 μm 1 kDa
14-558-250 63 mm Polysulfone 120 μm 10 kDa
14-558-256 25 mm Polysulfone 120 μm 30 kDa
14-558-261 25 mm Polysulfone 120 μm 300 kDa
14-558-252 25 mm Polysulfone 120 μm 50 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
32 options
Catalog No. 14-558-230 Supplier Sartorius Supplier No. 1443925D
Add to Cart
Add to Cart

For sample concentration in stirred cells

Sartorius Stedim Biotech offers a wide range of ultrafiltration membrane discs for sample concentration in stirred cells. Membrane disc diameters range from 13 mm to 293 mm, molecular weight cut-offs from 1 to 300 kDa. Different membrane materials are available depending on the application: PES is the standard material with high flux, no hydrophobic or hydrophilic interactions and low fouling characteristics.Hydrosart™, a patented regenerated cellulose membrane, features extremely low protein binding and extended chemical resistance and is preferred for concentration of samples with very low protein content.

  • Wide range of MWCO
  • Wide range of disc diameters
  • Compatible with all stirred cells
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Membrane Filter
Format Ultrafiltration Disc
Material Hydrosart
Material (Membrane) Cellulose
MWCO 10 kDa
Sterility Non-sterile
For Use With (Application) For sample concentration in stirred cells
Quantity 10/Pk.
Thickness (Metric) 180 μm
Type Membrane Filter
Diameter (Metric) 25 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.