Learn More
- Home
- Products
- Filters and Filtration
- Ultrafiltration Filters and Devices
- Sartorius Ultrafiltration Membrane Discs
Sartorius Ultrafiltration Membrane Discs
Catalog No. 14558257 Shop All Sartorius Products| Catalog No. | Diameter (Metric) | Material (Membrane) | Thickness (Metric) | MWCO |
|---|---|---|---|---|
| 14-558-257 | 47 mm | Polysulfone | 120 μm | 30 kDa |
| 14-558-242 | 25 mm | Polysulfone | 120 μm | 5 kDa |
| 14-558-255 | 76 mm | Polysulfone | 120 μm | 50 kDa |
| 14-558-225 | 25 mm | Cellulose | 180 μm | 5 kDa |
| 14-558-245 | 76 mm | Polysulfone | 120 μm | 5 kDa |
| 14-558-251 | 76 mm | Polysulfone | 120 μm | 10 kDa |
| 14-558-234 | 25 mm | Cellulose | 180 μm | 30 kDa |
| 14-558-244 | 63 mm | Polysulfone | 120 μm | 5 kDa |
| 14-558-228 | 63 mm | Cellulose | 180 μm | 5 kDa |
| 14-558-247 | 25 mm | Polysulfone | 120 μm | 10 kDa |
| 14-558-260 | 47 mm | PES | 120 μm | 100 kDa |
| 14-558-239 | 47 mm | Polysulfone | 120 μm | 1 kDa |
| 14-558-258 | 63 mm | Polysulfone | 120 μm | 30 kDa |
| 14-558-227 | 47 mm | Cellulose | 180 μm | 5 kDa |
| 14-558-259 | 76 mm | Polysulfone | 120 μm | 30 kDa |
| 14-558-246 | 150 mm | Polysulfone | 120 μm | 10 kDa |
| 14-558-253 | 47 mm | Polysulfone | 120 μm | 50 kDa |
| 14-558-237 | 76 mm | Cellulose | 180 μm | 30 kDa |
| 14-558-233 | 76 mm | Cellulose | 180 μm | 10 kDa |
| 14-558-235 | 47 mm | Cellulose | 180 μm | 30 kDa |
| 14-558-264 | 76 mm | Polysulfone | 120 μm | 300 kDa |
| 14-558-236 | 63 mm | Cellulose | 180 μm | 30 kDa |
| 14-558-240 | 63 mm | Polysulfone | 120 μm | 1 kDa |
| 14-558-249 | 47 mm | Polysulfone | 120 μm | 10 kDa |
| 14-558-229 | 76 mm | Cellulose | 180 μm | 5 kDa |
| 14-558-262 | 47 mm | Polysulfone | 120 μm | 300 kDa |
| 14-558-241 | 76 mm | Polysulfone | 120 μm | 1 kDa |
| 14-558-230 | 25 mm | Cellulose | 180 μm | 10 kDa |
| 14-558-250 | 63 mm | Polysulfone | 120 μm | 10 kDa |
| 14-558-256 | 25 mm | Polysulfone | 120 μm | 30 kDa |
| 14-558-261 | 25 mm | Polysulfone | 120 μm | 300 kDa |
| 14-558-252 | 25 mm | Polysulfone | 120 μm | 50 kDa |
Description
Sartorius Stedim Biotech offers a wide range of ultrafiltration membrane discs for sample concentration in stirred cells. Membrane disc diameters range from 13 mm to 293 mm, molecular weight cut-offs from 1 to 300 kDa. Different membrane materials are available depending on the application: PES is the standard material with high flux, no hydrophobic or hydrophilic interactions and low fouling characteristics.Hydrosart™, a patented regenerated cellulose membrane, features extremely low protein binding and extended chemical resistance and is preferred for concentration of samples with very low protein content.
- Wide range of MWCO
- Wide range of disc diameters
- Compatible with all stirred cells
Specifications
Specifications
| Filter Type | Membrane Filter |
| Format | Ultrafiltration Disc |
| Material | PES |
| Material (Membrane) | Polysulfone |
| MWCO | 30 kDa |
| Sterility | Non-sterile |
| For Use With (Application) | For sample concentration in stirred cells |
| Quantity | 10/Pk. |
| Thickness (Metric) | 120 μm |
| Type | Membrane Filter |
| Diameter (Metric) | 47 mm |
| Show More Show Less |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.