Learn More
Sigma Aldrich UltrafleXX TPE-E 3D printing filament
Catalog No. 503126058 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Ea.
Catalog No. 50-312-6058
Supplier: Sigma Aldrich TRD3D00511EA
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
ParametersExtruder (nozzle) temperature: 225 +/-10°CBed temperature: 45°C to 95°CPrint speed: 20-35 mm/sFan: 0-30 %Minimum nozzle diameter: 0.35 mmPlate materials: glass, blue tape, kapton tape, etc.
Specifications
Quantity 1 Ea.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.