Learn More
Innovative Med Tech UltraFlo-C Capillary Collector K2 EDTA
Description
- The ideal combination of controlled blood collection and sample container to avoid sample
- Exact measurement and stabilization of the required amount of blood via integrated end-to-end plastic capillary coated with anti-coagulant.
- Reduction of the infection risk thanks to unbreakable plastic capillaries.
- Clear marking of expiry date and production batch on each sample container
Specifications
Specifications
| Color | Purple |
| Product Type | Capillary Tube |
| Additive | K2 EDTA |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Closure Type | Snap-lock |
| Capacity (Metric) | 100 μL |