Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ultramarine Blue, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Ultramarine Blue
Specifications
Specifications
CAS Max % | 1 |
Percent Purity | 97 to 102% |
Grade | USP |
Packaging | Amber Glass Bottle |
Color | Colorless to Pale Yellow |
Quantity | 500 g |
Chemical Name or Material | Undecylenic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction