Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Branson Ultrasonics™ Ultrasonic General-Purpose Cleaning Solutions

Catalog No. p-2598663 Shop All Branson Ultrasonics Corporation Products
Change view
Click to view available options
Volume (English):
1 qt.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
23-260865 1 qt.
23-260866 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-260865 Supplier Branson Ultrasonics™ Supplier No. 000955014
Only null left
Add to Cart
Add to Cart

Safely clean metals including stainless-steel, tin, and copper

  • Safely and effectively remove soils, fingerprints, tissues and light oils from a variety of materials
  • Solutions contain no phosphates and are biodegradable
  • Dilute with water to the recommended working concentration listed on ordering table

Specifications

pH 11 to 12
Concentration 10 to 12%
For Use With (Application) Soils, fingerprints, tissue, light Oils
Volume (English) 1 qt.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.