Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Branson Ultrasonics™ Ultrasonic General-Purpose Cleaning Solutions
Click to view available options
Volume (English):
1 qt.
1 gal.
Description
- Safely and effectively remove soils, fingerprints, tissues and light oils from a variety of materials
- Solutions contain no phosphates and are biodegradable
- Dilute with water to the recommended working concentration listed on ordering table
Specifications
Specifications
pH | 11 to 12 |
Concentration | 10 to 12% |
For Use With (Application) | Soils, fingerprints, tissue, light Oils |
Volume (English) | 1 gal. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction