Learn More
United Scientific™ Ultrasonic Grating Apparatus
SDPSCI_ED
Catalog No. s23932
Click to view available options
:
Ultrasonic Grating Apparatus
Use the Ultrasonic Grating Apparatus to demonstrate the interaction of longitudinal sound waves and transverse light waves or precisely determine the speed of sound in liquids. Use with any clear non-aggressive liquid of low electrical conductivity.
  • The Ultrasonic Grating Apparatus is both a unique demonstration of the interaction of longitudinal sound waves and transverse light waves and also a method for the precise determination of the speed of sound in liquids, which is otherwise difficult to do in the undergraduate lab.
  • The core of the apparatus is a parallel-sided glass tank that houses the liquid sample and contains a lead zirconate titanate (PZT) ultrasonic transducer.
  • The transducer is energized by an RF signal in the 8 to 12 MHz range.
  • Resonances can be found to set up a standing wave field in the liquid, which results in a periodic variation of the liquid's refractive index along the tank.
Specifications
Material Glass
Product Type Optics Set
Shape Various
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.