Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UMOD Mouse anti-Human, FITC, Clone: 10.32, Abnova™

Catalog No. 89935079 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-935-079 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-935-079 Supplier Abnova Corporation Supplier No. MAB12564
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human UMOD.

This gene encodes uromodulin, the most abundant protein in normal urine. Its excretion in urine follows proteolytic cleavage of the ectodomain of its glycosyl phosphatidylinosital-anchored counterpart that is situated on the luminal cell surface of the loop of Henle. Uromodulin may act as a constitutive inhibitor of calcium crystallization in renal fluids. Excretion of uromodulin in urine may provide defense against urinary tract infections caused by uropathogenic bacteria. Defects in this gene are associated with the autosomal dominant renal disorders medullary cystic kidney disease-2 (MCKD2) and familial juvenile hyperuricemic nephropathy (FJHN). These disorders are characterized by juvenile onset of hyperuricemia, gout, and progressive renal failure. While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same isoform. [provided by RefSeq]

Specifications

Antigen UMOD
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone 10.32
Conjugate FITC
Description Mouse monoclonal antibody raised against human UMOD.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS (EIA grade BSA, 0.02% sodium azide).
Gene Alias ADMCKD2/FJHN/HNFJ/MCKD2/THGP/THP
Gene Symbols UMOD
Host Species Mouse
Immunogen Human UMOD
Purification Method Protein G purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7369
Target Species Human
Content And Storage Store in the dark at 4°C. For long term storage store at -80°C. Avoid prolonged exposure to light.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.