Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UMODL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598230
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598230 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598230 Supplier Invitrogen™ Supplier No. PA598230
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UMODL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

UMODL1 gene ontology annotations related to this gene include adipose tissue development; cellular response to gonadotropin-releasing hormone; multicellular organismal reproductive process; regulation of apoptotic process; regulation of gene expression; regulation of ovarian follicle development; single fertilization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UMODL1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene UMODL1
Gene Accession No. Q5DID0, Q5DID3
Gene Alias D17Ertd488e; Olfactorin; RGD1566039; Umodl1; uromodulin like 1; uromodulin-like 1
Gene Symbols UMODL1
Host Species Rabbit
Immunogen Recombinant Human Uromodulin-like 1 protein (537-654AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52020, 89766
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.