Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UMODL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564317 Supplier Invitrogen™ Supplier No. PA564317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSGRELCANL EGSYWCVCHQ EAPATSPRKL NLEWEDCPPV SDYVVLNVTS DSFQVSWRLN STQNHTFHVR VYRGMELLRS Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 59%.

UMODL1 gene ontology annotations related to this gene include adipose tissue development; cellular response to gonadotropin-releasing hormone; multicellular organismal reproductive process; regulation of apoptotic process; regulation of gene expression; regulation of ovarian follicle development; single fertilization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UMODL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UMODL1
Gene Accession No. Q5DID0
Gene Alias D17Ertd488e; Olfactorin; RGD1566039; Umodl1; uromodulin like 1; uromodulin-like 1
Gene Symbols UMODL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UMODL1. Recombinant protein control fragment (Product #RP-103651).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89766
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.