Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UMPS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112256
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112256 Supplier Invitrogen™ Supplier No. PA5112256
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UMPS Polyclonal Antibody for Western Blot, IHC (P), IP

This gene encodes a uridine 5'-monophosphate synthase. The encoded protein is a bifunctional enzyme that catalyzes the final two steps of the de novo pyrimidine biosynthetic pathway. The first reaction is carried out by the N-terminal enzyme orotate phosphoribosyltransferase which converts orotic acid to orotidine-5'-monophosphate. The terminal reaction is carried out by the C-terminal enzyme OMP decarboxylase which converts orotidine-5'-monophosphate to uridine monophosphate. Defects in this gene are the cause of hereditary orotic aciduria. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UMPS
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene UMPS
Gene Accession No. P11172
Gene Alias 1700095D23Rik; AA408257; AL033308; BB164745; mutant uridine monophosphate synthase; ODC; OK/SW-cl.21; OMPdecase; OPRT; OPRTase; orotate phosphoribosyl transferase and orotidine-5'-decarboxylase; orotate phosphoribosyltransferase; Orotidine 5'-phosphate decarboxylase; UMP synthase; UMPS; Uridine 5'-monophosphate synthase; uridine 5-monophosphate synthase; uridine 5'-monophosphate synthase-like protein; uridine monophophate synthase; uridine monophosphate synthase; uridine monophosphate synthetase; uridine monophosphate synthetase (orotate phosphoribosyl transferase and orotidine-5'-decarboxylase); uridine monophosphate synthetase [orotate phosphoribosyl transferase and orotidine-5'-decarboxylase]
Gene Symbols UMPS
Host Species Rabbit
Immunogen E. coli-derived Human UMPS fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7372
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.