Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ UMPS Recombinant Protein
SDP

Catalog No. NB6021320UG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB6021320UG 20 μg
NB60213100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB6021320UG Supplier Novus Biologicals™ Supplier No. NBP26021320UG
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-480 of Human UMPS The Recombinant Human UMPS Protein is derived from E. coli. The Recombinant Human UMPS Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation PBS (pH 7.4). containingno Preservative
Gene ID (Entrez) 7372
Name Recombinant UMPS Protein
Purification Method >95%, by SDS-PAGE
Quantity 20 μg
Source Baculovirus
Storage Requirements Store at 4°Cshort term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration < 1.0 EU per 1ug of protein (determined by LAL method)
Gene Alias EC 4.1.1.23, OMPdecase, OPRT, OPRTase, orotate phosphoribosyltransferase, orotidine 5'-phosphate decarboxylase, UMP synthase, uridine 5'-monophosphate synthase, uridine monophosphate synthetase
Gene Symbol UMPS
Product Type Recombinant Protein
Recombinant Recombinant
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.