Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UNC 0224
GREENER_CHOICE

Catalog No. 386110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-611-0 10 mg
38-615-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 38-611-0 Supplier Tocris Bioscience™ Supplier No. 3861/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent G9a and GLP inhibitor

Potent G9a and GLP inhibitor (IC50 values are 15 and 20 nM in a ThioGlo assay, respectively). Exhibits >1000-fold selectivity for G9a over SET7, SET8 and SET9.

Chemical Identifiers

CAS 1197196-48-7
Molecular Formula C26H43N7O2

Specifications

Quantity 10 mg
Formula Weight 485.67
Purity >98%
Molecular Weight (g/mol) 485.677
Chemical Name or Material UNC 0224
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.