Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UNC 0642
GREENER_CHOICE

Catalog No. 513210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-321-0 10 mg
51-325-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-321-0 Supplier Tocris Bioscience™ Supplier No. 5132/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective G9a and GLP inhibitor

UNC 0642 is a potent and selective G9a and GLP histone lysine methyltransferase inhibitor (IC50 < 2.5 nM) that exhibits >2,000-fold selectivity for G9a and GLP over PRC2-EZH2 and >20,000-fold selectivity over other methyltransferases. UNC 0642 reduces H3K9 dimethylation levels in MDA-MB-231 cells (IC50 = 110 nM), and displays modest brain penetration in vivo.

Specifications

Quantity 10 mg
Product Type UNC 0642
Purity >99%
Molecular Weight (g/mol) 546.7
Inhibitors UNC 0642

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.