Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UNC 1999
GREENER_CHOICE

Catalog No. 490410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-041-0 10 mg
49-045-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-041-0 Supplier Tocris Bioscience™ Supplier No. 4904/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective EZH2/EZH1 inhibitor

UNC 1999 is a potent and SAM-competitive EZH2/EZH1 lysine methyltransferase inhibitor (IC50 values are 10 and 45 nM, respectively). Selective for EZH2/EZH1 over a panel of other methyltransferases and non-epigenetic targets. Reduces H3K27me3 levels in vitro. Prolongs survival of MLL-AF9 bearing mice. Orally bioavailable.

Negative Control also available.

Specifications

Quantity 10 mg
Product Type UNC 1999
Purity >99%
Molecular Weight (g/mol) 569.74
Inhibitors UNC 1999

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.