Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ UNC 3230
GREENER_CHOICE

Catalog No. 527110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-711-0 10 mg
52-715-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-711-0 Supplier Tocris Bioscience™ Supplier No. 5271/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PIP5K1C inhibitor; antinociceptive

UNC 3230 is a potent and selective PIP5K1C inhibitor (IC50 = 41 nM). Exhibits selectivity for PIP5K1C over PIP5K1A, the PI 3-kinase family and a panel of other kinases. Reduces PIP2 levels and LPA-induced calcium signaling in dorsal root ganglia (DRG) neurons in vitro. Reduces nociception in mouse models of chronic pain.

Specifications

Quantity 10 mg
Product Type UNC 3230
Purity >99%
Molecular Weight (g/mol) 344.43
Inhibitors UNC 3230

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.