Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC119 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728461 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8461 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8461 Supplier Proteintech Group Inc Supplier No. 130651AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is specifically expressed in the photoreceptors in the retina. The encoded product shares strong homology with the C. elegans unc119 protein and it can functionally complement the C. elegans unc119 mutation. It has been localized to the photoreceptor synapses in the outer plexiform layer of the retina, and suggested to play a role in the mechanism of photoreceptor neurotransmitter release through the synaptic vesicle cycle. Two transcript variants encoding different isoforms have been described for this gene.

Specifications

Antigen UNC119
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene UNC119
Gene Accession No. O19177, Q13432, Q62885, Q9Z2R6
Gene Alias HRG4, Protein unc 119 homolog A, Retinal protein 4, RG4, unc 119 homolog (C. elegans), UNC119
Gene Symbols UNC119
Host Species Rabbit
Immunogen UNC119 Fusion Protein Ag3757
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22248, 29402, 491169, 9094
Target Species Canine, Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.