Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC119B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140033 Supplier Invitrogen™ Supplier No. PA5140033
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: IRDLETGTVL FEIAKPCVSD QEEDEEEGGG DVDISAGR Highest antigen sequence indentity to the following orthologs: Mouse - 79%, Rat - 79%.

Mouse Unc119b is related closely to Unc119, a homolog of C. elegans unc119 protein which was first discovered in C. elegans on the basis of a spontaneous mutation affecting locomotion, feeding behavior and chemosensation. Mouse Unc119b shares 90% sequence identity with human UNC119b protein. UNC119b is a myristoyl-binding protein that acts as a cargo adapter: specifically binds the myristoyl moiety of a subset of N-terminally myristoylated proteins and is required for their localization. Myristoyl-binding activity of UNC119b is required to localize NPHP3 to the primary cilium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC119B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UNC119B
Gene Accession No. A6NIH7
Gene Alias AA407659; AI414892; POC7 centriolar protein homolog B; POC7B; Protein unc-119 homolog B; protein unc-119 homolog B; LOW QUALITY PROTEIN: protein unc-119 homolog B; unc119 homolog B; unc-119 homolog B; unc-119 homolog B (C. elegans); unc-119 lipid binding chaperone B; UNC119B
Gene Symbols UNC119B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UNC119B. Recombinant protein control fragment (Product #RP-109649).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84747
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.