Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UNC13B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555091
Description
Immunogen sequence: SQLSELDQYH EQDDDHRETD SIHSCHSSHS LSRDGQAGFG EQEKPLEVTG QAEKEAACEP KEMKEDATTH PP Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 69%.
Plays a role in vesicle maturation during exocytosis as a target of the diacylglycerol second messenger pathway. Is involved in neurotransmitter release by acting in synaptic vesicle priming prior to vesicle fusion and participates in the activity-depending refilling of readily releasable vesicle pool (RRP). Essential for synaptic vesicle maturation in a subset of excitatory/glutamatergic but not inhibitory/GABA-mediated synapses.
Specifications
UNC13B | |
Polyclonal | |
Unconjugated | |
UNC13B | |
homolog of rat Munc13 (diacylglycerol-binding); munc13; Munc13-1; Munc13-2; protein unc-13 homolog B; UNC13; unc13 homolog 1; UNC-13 homolog 1; unc-13 homolog A; unc-13 homolog B; unc-13 homolog B (C. elegans); Unc13a; Unc13b; Unc13h1; Unc13h2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10497 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14795 | |
UNC13B | |
Recombinant protein corresponding to Human UNC13B. Recombinant protein control fragment (Product #RP-93102). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction