Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC13B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590699
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590699 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590699 Supplier Invitrogen™ Supplier No. PA590699
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSLLCVRVKR AKFQGSPDKF NTYVTLKVQN VKSTTVAVRG DQPSWEQDFM FEISRLDLGL SVEVWNKGLI WDTMVGTVWI ALKTIRQSDE EGPGEWSTLE AETLMKDDEI CGTRNPTPHK ILLDTRFELP FDIPEEEARY WTYKWEQINA LGADNEYSSQ EESQRKPLPT AAAQCSFEDP DSAVDDRDSD YRSETSNSFP PPYHTASQPN ASVHQFPVPV RSPQQLLLQG SSRDSCNDSM QSYDLDYPER RAISPTSSSR YGSSCNVSQG SSQLSELDQY HEQDDDHRET DSIHSCHSSH SLSRDGQAGF GEQEKPLEVT GQAEKEAACE PKEMKEDATT HPPPDLVLQK; Positive Samples: 293T, LO2; Cellular Location: Cell junction, Cell membrane, Cytoplasm, Membrane, Peripheral membrane protein, synapse.

Plays a role in vesicle maturation during exocytosis as a target of the diacylglycerol second messenger pathway. Is involved in neurotransmitter release by acting in synaptic vesicle priming prior to vesicle fusion and participates in the activity-depending refilling of readily releasable vesicle pool (RRP). Essential for synaptic vesicle maturation in a subset of excitatory/glutamatergic but not inhibitory/GABA-mediated synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC13B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene UNC13B
Gene Accession No. O14795, Q9Z1N9
Gene Alias homolog of rat Munc13 (diacylglycerol-binding); munc13; Munc13-1; Munc13-2; protein unc-13 homolog B; UNC13; unc13 homolog 1; UNC-13 homolog 1; unc-13 homolog A; unc-13 homolog B; unc-13 homolog B (C. elegans); Unc13a; Unc13b; Unc13h1; Unc13h2
Gene Symbols UNC13B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human UNC13B (NP_0063683).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10497, 22249
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.