Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNC13B Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572594
Description
Plays a role in vesicle maturation during exocytosis as a target of the diacylglycerol second messenger pathway. Is involved in neurotransmitter release by acting in synaptic vesicle priming prior to vesicle fusion and participates in the activity-depending refilling of readily releasable vesicle pool (RRP). Essential for synaptic vesicle maturation in a subset of excitatory/glutamatergic but not inhibitory/GABA-mediated synapses (By similarity).
Specifications
UNC13B | |
Polyclonal | |
Unconjugated | |
UNC13B | |
MUNC13; UNC13; Unc13h2; homolog of rat Munc13 (diacylglycerol-binding); munc13-2; protein unc-13 homolog B | |
Rabbit | |
Protein A | |
RUO | |
10497 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.66 mg/mL | |
PBS with 0.09% sodium azide | |
O14795 | |
UNC13B | |
KLH conjugated synthetic peptide between 1062-1091 amino acids from the Central region of human UNC13B | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction