Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNC13D/Munc 13-4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001434
Description
UNC13D/Munc 13-4 Polyclonal specifically detects UNC13D/Munc 13-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
UNC13D/Munc 13-4 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FHL3, HLH3, HPLH3, MUNC13-4, Munc13-4protein unc-13 homolog D, unc-13 homolog D (C. elegans) | |
Goat | |
Affinity Purified | |
RUO | |
201294 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin 5ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_954712. | |
UNC13D | |
Peptide with sequence C-KQASQHALRPAP corresponding to C-Terminus according to NP_954712.1. | |
0.1 mg | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction