Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Unc18-2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550281
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550281 Supplier Invitrogen™ Supplier No. PA550281
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total STXBP2 protein.

This gene encodes a member of the STXBP/unc-18/SEC1 family. The encoded protein is involved in intracellular trafficking, control of SNARE (soluble NSF attachment protein receptor) complex assembly, and the release of cytotoxic granules by natural killer cells. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Unc18-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene STXBP2
Gene Accession No. Q15833
Gene Alias C79054; FHL5; Hunc18b; Munc-18-2; Munc18-2; Munc18b; Munc-18b; muSec1; pp10122; Protein unc-18 homolog 2; protein unc-18 homolog B; Stxbp2; Sxtbp2; Sxtp2; syntaxin binding protein 2; syntaxin binding protein Munc18-2; syntaxin-binding protein 2; unc-18 homolog 2; unc-18 homolog B; Unc18-2; Unc18b; unc-18B
Gene Symbols STXBP2
Host Species Rabbit
Immunogen Fusion protein corresponding to residues near the C terminal of human Syntaxin binding protein 2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6813
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.