Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Unc18-3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987278
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7278 Supplier Bioss Supplier No. BS6462R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen Unc18-3
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene STXBP3
Gene Accession No. O00186
Gene Alias Mammalian homolog of Unc-18c; MUNC-18-3; MUNC18-3; MUNC18C; Munc-18c; Munc18-c; Platelet Sec1 protein; Protein unc-18 homolog 3; Protein unc-18 homolog C; PSP; Stxbp3; Stxbp3a; Sxtbp3; syntaxin 4 binding protein; syntaxin binding protein 3; syntaxin binding protein 3A; Syntaxin-binding protein 3; Syntaxin-BP3; unc-18 homolog 3; Unc18-3; Unc18c; Unc-18C
Gene Symbols STXBP3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Unc18-3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6814
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.