Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNC45A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501732457 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2457 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2457 Supplier Proteintech Group Inc Supplier No. 195641AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody recognizes the C-term of UNC45A.

UNC45A, also named as SMAP1, acts as co-chaperone for HSP90. UNC45A prevents the stimulation of HSP90AB1 ATPase activity by AHSA1. It is a positive factor in promoting PGR function in the cell. It is necessary for proper folding of myosin and necessary for normal cell proliferation, normal myotube formation and myosin accumulation during muscle cell development. UNC45A plays a role in erythropoiesis in stroma cells in the spleen.

Specifications

Antigen UNC45A
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene UNC45A
Gene Accession No. Q32PZ3, Q99KD5, Q9H3U1
Gene Alias GC UNC45, GCUNC 45, GCUNC45, IRO039700, Protein unc 45 homolog A, SMAP 1, SMAP1, unc 45 homolog A (C. elegans), Unc 45A, UNC45A
Gene Symbols UNC45A
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101869, 308759, 55898
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.