Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC45B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518748
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518748 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518748 Supplier Invitrogen™ Supplier No. PA518748
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.

UNC45B plays a role in myoblast fusion and sarcomere organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC45B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Unc45b
Gene Accession No. Q8IWX7
Gene Alias AA445617; cardiomyopathy associated 4; CMYA4; CTRCT43; D230041A13Rik; FLJ38610; MGC119540; MGC119541; Protein unc-45 homolog B; SMUNC45; striated muscle UNC45; UNC45; unc-45 homolog B; unc-45 homolog B (C. elegans); unc-45 myosin chaperone B; Unc45b; UNC-45B
Gene Symbols Unc45b
Host Species Goat
Immunogen Synthetic peptide sequence (RSDKLRQKIFKER) corresponding to the internal amino acids of UNC45B (aa 749-762).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146862
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.