Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC45B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571870
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571870 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571870 Supplier Invitrogen™ Supplier No. PA571870
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UNC45B Polyclonal Antibody for Western Blot, IHC (P)

UNC45B plays a role in myoblast fusion and sarcomere organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC45B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Unc45b
Gene Accession No. Q8CGY6, Q8IWX7
Gene Alias AA445617; cardiomyopathy associated 4; CMYA4; CTRCT43; D230041A13Rik; FLJ38610; MGC119540; MGC119541; Protein unc-45 homolog B; SMUNC45; striated muscle UNC45; UNC45; unc-45 homolog B; unc-45 homolog B (C. elegans); unc-45 myosin chaperone B; Unc45b; UNC-45B
Gene Symbols Unc45b
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 235-264 amino acids from the N-terminal region of human UNC45B.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146862, 217012
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.