Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNC5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547133
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547133 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547133 Supplier Invitrogen™ Supplier No. PA547133
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, less than 5% cross-reactivity with recombinant rat UNC5H2, recombinant human (rh) UNC5H3 and rhUNC5H4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

The UNC5H family of proteins act as transmembrane receptors for netrin-1 and play a crucial role in axon guidance and migration of neural cells. Additionally, when cleaved by a caspase to produce an intracellular fragment containing a death domain, UNC5H receptors induce apoptosis. This activity is blocked by the binding of netrin-1. In the absence of netrin-1, UNC5H receptors act as tumor suppressors by inhibiting anchorage-independent growth and invasion, but mutation of these receptors provides a potential mechanism for tumorigenicity. The expression of UNC5H receptors is downregulated in multiple carcinomas, including colorectal, breast, ovary, uterus, stomach, lung and kidney cancers. UNC5H1, also known as UNC5HA (unc-5 homolog A), is a member of the UNC5H family of proteins and is localized to the cell membrane. Three isoforms of UNC5H1 exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNC5A
Applications Neutralization, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Unc5a
Gene Accession No. O08721
Gene Alias Kiaa1976; mKIAA1976; netrin receptor UNC5A; netrin receptor Unc5h1; protein unc-5 homolog 1; Protein unc-5 homolog A; transmembrane receptor Unc5H1; unc5 (C.elegans homolog) a; unc5 homolog 1; unc-5 homolog 1; unc-5 homolog A; unc-5 homolog A (C. elegans); unc-5 netrin receptor A; UNC5A; Unc5h1; UNC5H1FLJ16449
Gene Symbols Unc5a
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant rat UNC5H1 Gln26-Asp358.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60629
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.