Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UNC5B Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA537993
Description
This antibody is predicted to react with mouse, rat, dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Receptor for netrin required for axon guidance. Mediates axon repulsion of neuronal growth cones in the developing nervous system upon ligand binding. Axon repulsion in growth cones may be caused by its association with DCC that may trigger signaling for repulsion. It also acts as a dependence receptor required for apoptosis induction when not associated with netrin ligand. Mediates apoptosis by activating DAPK1. In the absence of NTN1, activates DAPK1 by reducing its autoinhibitory phosphorylation at Ser-308 thereby increasing its catalytic activity.
Specifications
UNC5B | |
Polyclonal | |
Unconjugated | |
UNC5B | |
6330415E02Rik; A630020F16; C. elegans; D10Bwg0792e; Netrin receptor UNC5B; p53RDL1; p53-regulated receptor for death and life protein 1; Protein unc-5 homolog 2; protein unc-5 homolog B; transmembrane receptor Unc5H2; unc5 homolog 2; unc-5 homolog 2; unc-5 homolog B; unc-5 homolog B (C. elegans); unc-5 netrin receptor B; Unc5b; UNC5H2; UNQ1883/PRO4326 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
219699 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8IZJ1 | |
UNC5B | |
Peptide with sequence C-SARDWIFQLKTQAHQ, from the internal region of UNC5B (aa629-643). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction